Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Unc5d
- Entrez Gene ID
-
210801
- Gene Name
- unc-5 homolog D (C. elegans)
- Gene Aliases
-
D930029E11Rik, Unc5h4, mKIAA1777
- Chromosome Location
-
Chr.8: 28646717 - 29219682 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC145695.1 |
- |
6 - 7 |
1000 |
71 |
View Full Product Details