Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exoc1
Entrez Gene ID
69940
Gene Name
exocyst complex component 1
Gene Aliases
2810407P21Rik, A730011E05Rik, SEC3, Sec3l1, Sec3p
Chromosome Location
Chr.5: 76529311 - 76570298 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289770.1 NP_001276699.1 9 - 10 1379 130
NM_001289771.1 NP_001276700.1 9 - 10 1379 130
NM_027270.2 NP_081546.1 10 - 11 1400 130
GenBank mRNA AK155980.1 - 9 - 10 1333 130
BC024678.1 - 9 - 10 1276 130
BC029330.1 - 2 - 3 228 130
BC064811.1 - 10 - 11 1388 130
BC087543.1 - 9 - 10 1315 130

View Full Product Details