Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zar1
Entrez Gene ID
317755
Gene Name
zygote arrest 1
Gene Aliases
-
Chromosome Location
Chr.5: 72577113 - 72581361 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174877.3 NP_777366.1 2 - 3 890 65
GenBank mRNA AY191415.1 - 2 - 3 890 65
BC099399.1 - 2 - 3 884 65

View Full Product Details