The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195496.1 | NP_001182425.1 | 10 - 11 | 2182 | 73 |
NM_001195497.1 | NP_001182426.1 | 9 - 10 | 2134 | 73 | |
NM_001195498.1 | NP_001182427.1 | 10 - 11 | 2185 | 73 | |
NM_001195499.1 | NP_001182428.1 | 10 - 11 | 2182 | 73 | |
NM_001195500.1 | NP_001182429.1 | 9 - 10 | 2134 | 73 | |
NM_027539.5 | NP_081815.3 | 9 - 10 | 2137 | 73 | |
GenBank mRNA | AB198721.1 | - | 10 - 11 | 1539 | 73 |
AK049126.1 | - | 9 - 10 | 1624 | 73 | |
AK049179.1 | - | 9 - 10 | 1586 | 73 | |
AK082633.1 | - | 9 - 10 | 1633 | 73 | |
AY968049.1 | - | 10 - 11 | 1470 | 73 | |
AY968050.1 | - | 10 - 11 | 1473 | 73 | |
AY968051.1 | - | 10 - 11 | 1470 | 73 | |
BC052071.1 | - | 2 - 3 | 178 | 73 | |
BC056921.1 | - | 9 - 10 | 2105 | 73 | |
BC060192.1 | - | 9 - 10 | 1438 | 73 |