Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rps6ka2
Entrez Gene ID
20112
Gene Name
ribosomal protein S6 kinase, polypeptide 2
Gene Aliases
90kDa, D17Wsu134e, Rps6ka-rs1, Rsk3, p90rsk, pp90rsk
Chromosome Location
Chr.17: 7088975 - 7303315 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011299.4 NP_035429.1 2 - 3 447 84
GenBank mRNA AJ131021.1 - 2 - 3 275 84
AK132079.1 - 1 - 2 115 84
BC043064.1 - 2 - 3 397 84
BC051079.1 - 2 - 3 357 84
BC055331.1 - 2 - 3 447 84
BC056946.1 - 2 - 3 390 84

View Full Product Details