Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pik3cb
Entrez Gene ID
74769
Gene Name
phosphatidylinositol 3-kinase, catalytic, beta polypeptide
Gene Aliases
1110001J02Rik, AI447572, p110beta
Chromosome Location
Chr.9: 99038401 - 99140211 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029094.3 NP_083370.2 21 - 22 3128 105
GenBank mRNA AK003230.1 - 5 - 6 543 105
AK081350.1 - 21 - 22 3319 105
AK090116.1 - 21 - 22 3005 105
AK154106.1 - 21 - 22 3113 105
AK165230.1 - - 588 105
BC127073.1 - 20 - 21 2777 105

View Full Product Details