Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hcfc2
Entrez Gene ID
67933
Gene Name
host cell factor C2
Gene Aliases
1700129L13Rik, AU019838
Chromosome Location
Chr.10: 82697497 - 82795865 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081218.1 NP_001074687.1 6 - 7 1008 105

View Full Product Details