Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Unc50
Entrez Gene ID
67387
Gene Name
unc-50 homolog (C. elegans)
Gene Aliases
1110002A21Rik, GMH1, UNCL, URP
Chromosome Location
Chr.1: 37430144 - 37439124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK003273.1 - 1 - 2 93 89
AK004433.1 - 1 - 2 82 89

View Full Product Details