Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wac
Entrez Gene ID
225131
Gene Name
WW domain containing adaptor with coiled-coil
Gene Aliases
1110067P07Rik, A230035H12Rik, AI256735, AI256776, Wwp4
Chromosome Location
Chr.18: 7868459 - 7929028 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282093.1 NP_001269022.1 6 - 7 831 107
NM_153085.4 NP_694725.3 6 - 7 990 107
GenBank mRNA AF320996.1 - 6 - 7 770 107
AK156780.1 - 6 - 7 815 107
AK172416.1 - 6 - 7 814 107
BC057178.1 - 6 - 7 623 107
BC080851.1 - 6 - 7 948 107
CN668018.1 - 2 - 3 191 107

View Full Product Details