Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Armc4
Entrez Gene ID
74934
Gene Name
armadillo repeat containing 4
Gene Aliases
4930463I21Rik, b2b227.1Clo, b2b643Clo
Chromosome Location
Chr.18: 7088233 - 7301128 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081393.1 NP_001074862.1 13 - 14 1863 71
GenBank mRNA BC158097.1 - 12 - 13 1852 71

View Full Product Details