Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dzip1
Entrez Gene ID
66573
Gene Name
DAZ interacting protein 1
Gene Aliases
2510025K24Rik, 2810422M04Rik
Chromosome Location
Chr.14: 118875520 - 118925520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK013136.1 - 7 - 8 672 145
AK032834.1 - 13 - 14 1281 145
AK129256.1 - 16 - 17 2082 145
AK141247.1 - 18 - 19 2498 145
BC051931.1 - 4 - 5 480 145
BC060196.1 - 13 - 14 2839 145
BC098211.1 - 18 - 19 2570 145

View Full Product Details