The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029561.4 | NP_083837.2 | 7 - 8 | 1026 | 71 |
GenBank mRNA | AK002978.1 | - | 5 - 6 | 545 | 71 |
AK031073.1 | - | 7 - 8 | 841 | 71 | |
AK049678.1 | - | 7 - 8 | 896 | 71 | |
AK078939.1 | - | 7 - 8 | 936 | 71 | |
AK132235.1 | - | 7 - 8 | 955 | 71 | |
AK150832.1 | - | 7 - 8 | 840 | 71 | |
BC071237.1 | - | 7 - 8 | 945 | 71 |