The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015507.2 | NP_001015507.1 | 17 - 18 | 3722 | 64 |
GenBank mRNA | AK122372.1 | - | 17 - 18 | 3685 | 64 |
BC065119.1 | - | 17 - 18 | 3722 | 64 |