Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vprbp
Entrez Gene ID
321006
Gene Name
Vpr (HIV-1) binding protein
Gene Aliases
AI447437, B930007L02Rik, mKIAA0800
Chromosome Location
Chr.9: 106814172 - 106880992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015507.2 NP_001015507.1 8 - 9 1251 99
GenBank mRNA AK122372.1 - 8 - 9 1214 99
BC065119.1 - 8 - 9 1251 99

View Full Product Details