Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eml4
Entrez Gene ID
78798
Gene Name
echinoderm microtubule associated protein like 4
Gene Aliases
4930443C24Rik, AI644019
Chromosome Location
Chr.17: 83350931 - 83480361 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114361.1 NP_001107833.1 23 - 24 2788 81
NM_001114362.1 NP_001107834.1 21 - 22 2581 81
NM_001286567.1 NP_001273496.1 23 - 24 2691 81
NM_199466.3 NP_955760.3 20 - 21 2452 81
GenBank mRNA AK139067.1 - 6 - 7 1161 81
AK144604.1 - 23 - 24 2789 81
AK168655.1 - 1 - 2 86 81
AK168681.1 - 1 - 2 86 81
BC032258.1 - 9 - 10 936 81
BC065178.1 - 23 - 24 2629 81
BC066099.1 - 20 - 21 2383 81
BC067011.1 - 20 - 21 2401 81
BC070427.1 - 21 - 22 2521 81
BC079619.1 - 20 - 21 2383 81
DQ230326.1 - 22 - 23 2471 81

View Full Product Details