Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr74
Entrez Gene ID
107071
Gene Name
WD repeat domain 74
Gene Aliases
5730436H21Rik, AA407588
Chromosome Location
Chr.19: 8735799 - 8740624 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134139.1 NP_598900.1 4 - 5 372 70
GenBank mRNA AK077521.1 - 4 - 5 373 70
BC019968.1 - 4 - 5 372 70

View Full Product Details