The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037744.1 | NP_001032833.1 | 1 - 2 | 345 | 76 |
GenBank mRNA | BC099499.1 | - | 1 - 2 | 345 | 76 |