Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir17hg
- Entrez Gene ID
-
75957
- Gene Name
- Mir17 host gene (non-protein coding)
- Gene Aliases
-
5033413D16Rik, Mirhg1, OTTMUSG00000033261
- Chromosome Location
-
Chr.14: 115044390 - 115046728 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029382.1 |
|
1 - 1 |
|
130 |
GenBank mRNA |
AK053349.1 |
- |
1 - 1 |
|
130 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE332918.1 |
- |
1 |
|
|
Chr. 14 - 115045191 - 115045686 [+] on Build GRCm38 |
130 |
GenBankmRNA |
BF019638.1 |
- |
1 |
|
|
Chr. 14 - 115045321 - 115045741 [+] on Build GRCm38 |
130 |
GenBankmRNA |
AA174312.1 |
- |
1 |
|
|
Chr. 14 - 115045032 - 115045587 [+] on Build GRCm38 |
130 |
GenBankmRNA |
CD548669.1 |
- |
1 |
|
|
Chr. 14 - 115045027 - 115045620 [+] on Build GRCm38 |
130 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 92-1 |
Mir92-1 |
Mirn92-1, mir-92a-1 |
-521.0
|
Sense |
Chr. - 115044427 - 115044506 on Build GRCm38 |
Downstream |
glypican 5 |
Gpc5 |
A230034F01Rik |
46.6kb
|
Sense |
Chr. - 115092212 - 116525192 on Build GRCm38 |
View Full Product Details