The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019742.4 | NP_062716.1 | 2 - 3 | 396 | 113 |
GenBank mRNA | AF123387.1 | - | 2 - 3 | 283 | 113 |
AK004430.1 | - | 2 - 3 | 356 | 113 | |
AK082684.1 | - | 2 - 3 | 397 | 113 | |
BC048477.1 | - | 2 - 3 | 396 | 113 |