Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ywhaq
Entrez Gene ID
22630
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide
Gene Aliases
2700028P07Rik, AA409740, AU021156, R74690
Chromosome Location
Chr.12: 21390329 - 21417436 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011739.3 NP_035869.1 2 - 3 770 80
GenBank mRNA AK008948.1 - 3 - 4 540 80
AK012540.1 - 3 - 4 505 80
AK145568.1 - 3 - 4 495 80
AK146371.1 - 3 - 4 495 80
AK150850.1 - 3 - 4 495 80
AK151437.1 - 3 - 4 544 80
AK151715.1 - 3 - 4 523 80
AK151864.1 - 3 - 4 544 80
AK159403.1 - 3 - 4 528 80
AK159817.1 - 3 - 4 544 80
AK163237.1 - 3 - 4 497 80
AK167397.1 - 3 - 4 496 80
BC003837.1 - 2 - 3 283 80
BC080802.1 - 3 - 4 485 80
BC085299.1 - 3 - 4 550 80
BC090838.1 - 2 - 3 770 80
BC106164.1 - 2 - 3 472 80
BC110680.1 - 2 - 3 479 80
BC132252.1 - 2 - 3 539 80
BC132530.1 - 2 - 3 539 80
D87662.1 - 2 - 3 462 80
U56243.1 - 2 - 3 445 80
U57312.1 - 2 - 3 425 80

View Full Product Details