Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc29a2
Entrez Gene ID
13340
Gene Name
solute carrier family 29 (nucleoside transporters), member 2
Gene Aliases
Der12, Ent2, Hnp36
Chromosome Location
Chr.19: 5023407 - 5031972 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007854.3 NP_031880.2 9 - 10 1210 72
GenBank mRNA AK010950.1 - 9 - 10 1147 73
AK083484.1 - 9 - 10 1107 72
AK144451.1 - 9 - 10 1103 72
AK154310.1 - 9 - 10 1145 72
BC048958.1 - 9 - 10 1356 72
X86682.1 - 7 - 8 778 72

View Full Product Details