The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019807.2 | NP_062781.2 | 7 - 8 | 845 | 143 |
NM_207668.2 | NP_997551.1 | 7 - 8 | 845 | 143 | |
GenBank mRNA | AF210243.1 | - | 7 - 8 | 798 | 143 |
AK020693.1 | - | 7 - 8 | 807 | 143 | |
AK029273.1 | - | 7 - 8 | 831 | 143 | |
BC139826.1 | - | 7 - 8 | 836 | 143 |