The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019807.2 | NP_062781.2 | 9 - 10 | 1032 | 71 |
NM_207668.2 | NP_997551.1 | 9 - 10 | 1032 | 71 | |
GenBank mRNA | AF210243.1 | - | 9 - 10 | 985 | 71 |
AK029273.1 | - | 9 - 10 | 1018 | 71 | |
AK076383.1 | - | 9 - 10 | 1039 | 71 | |
BC139826.1 | - | 9 - 10 | 1023 | 71 |