Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nmnat3
- Entrez Gene ID
-
74080
- Gene Name
- nicotinamide nucleotide adenylyltransferase 3
- Gene Aliases
-
4933408N02Rik, PNAT3
- Chromosome Location
-
Chr.9: 98287432 - 98420442 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK041526.1 |
- |
4 - 5 |
543 |
75 |
View Full Product Details