The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133771.2 | NP_598532.1 | 8 - 9 | 876 | 103 |
GenBank mRNA | AF363447.1 | - | 9 - 10 | 938 | 103 |
AK002639.1 | - | 3 - 4 | 281 | 103 | |
AK159560.1 | - | 8 - 9 | 855 | 103 | |
BC013819.1 | - | 8 - 9 | 893 | 103 |