Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ndc80
- Entrez Gene ID
-
67052
- Gene Name
- NDC80 homolog, kinetochore complex component (S. cerevisiae)
- Gene Aliases
-
2610020P18Rik, AV139730, HEC, HEC1, Kntc2
- Chromosome Location
-
Chr.17: 71496100 - 71526857 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK132008.1 |
- |
4 - 5 |
322 |
82 |
View Full Product Details