Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgif1
Entrez Gene ID
21815
Gene Name
TGFB-induced factor homeobox 1
Gene Aliases
AA959811, AI462167, Tgif
Chromosome Location
Chr.17: 70844205 - 70853532 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164075.1 NP_001157547.1 1 - 2 429 60

View Full Product Details