The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025572.1 | NP_001020743.1 | 12 - 13 | 6154 | 69 |
GenBank mRNA | AK045925.1 | - | 4 - 5 | 1189 | 69 |
AK122392.1 | - | 4 - 5 | 2515 | 69 | |
BC028980.1 | - | 4 - 5 | 524 | 69 |