Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnot2
Entrez Gene ID
72068
Gene Name
CCR4-NOT transcription complex, subunit 2
Gene Aliases
2600016M12Rik, 2810470K03Rik, AA537049, AA959607, AW557563, C79650
Chromosome Location
Chr.10: 116485161 - 116581535 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037846.3 NP_001032935.2 2 - 3 206 136
NM_001037847.2 NP_001032936.2 2 - 3 136 136
NM_001037848.3 NP_001032937.1 2 - 3 206 136
GenBank mRNA AK078121.1 - 2 - 3 196 136
AK149767.1 - 2 - 3 180 136
AK167834.1 - 2 - 3 184 136
AK172132.1 - 2 - 3 193 136
BC065171.1 - 2 - 3 136 136

View Full Product Details