The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160097.1 | NP_001153569.1 | 1 - 2 | 506 | 69 |
NM_023878.3 | NP_076367.2 | 1 - 2 | 506 | 69 | |
GenBank mRNA | AK020131.1 | - | 1 - 2 | 470 | 69 |
BC021770.1 | - | 1 - 2 | 459 | 69 | |
CA467408.1 | - | - | 543 | 69 |