The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163622.1 | NP_001157094.1 | 12 - 13 | 2377 | 70 |
NM_001163623.1 | NP_001157095.1 | 11 - 12 | 1841 | 70 | |
NM_001163624.1 | NP_001157096.1 | 11 - 12 | 1709 | 70 | |
NM_145984.3 | NP_666096.3 | 12 - 13 | 1941 | 70 | |
GenBank mRNA | AK028877.1 | - | 12 - 13 | 2335 | 70 |
AK050878.1 | - | 11 - 12 | 1703 | 70 | |
AK083070.1 | - | 12 - 13 | 1882 | 70 | |
AK129142.1 | - | 6 - 7 | 1795 | 70 | |
BC004612.1 | - | 11 - 12 | 1795 | 70 |