The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163622.1 | NP_001157094.1 | 3 - 4 | 813 | 85 |
NM_001163623.1 | NP_001157095.1 | 2 - 3 | 277 | 85 | |
NM_001163624.1 | NP_001157096.1 | 2 - 3 | 145 | 85 | |
NM_145984.3 | NP_666096.3 | 3 - 4 | 377 | 85 | |
GenBank mRNA | AK028877.1 | - | 3 - 4 | 771 | 85 |
AK050878.1 | - | 2 - 3 | 139 | 85 | |
AK083070.1 | - | 3 - 4 | 318 | 85 | |
AK163108.1 | - | 3 - 4 | 778 | 85 | |
BC004612.1 | - | 2 - 3 | 231 | 85 | |
BC129880.1 | - | 1 - 2 | 809 | 85 | |
BY257815.1 | - | 2 - 3 | 146 | 85 |