Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alx1
Entrez Gene ID
216285
Gene Name
ALX homeobox 1
Gene Aliases
AI314867, C130005I02, Cart1
Chromosome Location
Chr.10: 103007847 - 103029452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172553.4 NP_766141.1 1 - 2 397 69
GenBank mRNA AK047846.1 - 1 - 2 398 69
AK086731.1 - 1 - 2 210 69
BC052200.1 - 1 - 2 379 69

View Full Product Details