Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fermt3
Entrez Gene ID
108101
Gene Name
fermitin family homolog 3 (Drosophila)
Gene Aliases
C79673, Kindlin3
Chromosome Location
Chr.19: 6998958 - 7019469 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153795.2 NP_722490.1 6 - 7 996 91
GenBank mRNA AK142912.1 - 6 - 7 997 91
AK158357.1 - 6 - 7 893 92
AK169682.1 - 6 - 7 907 91
BC025119.1 - 6 - 7 928 91
BC032204.1 - 6 - 7 972 91
BC066803.1 - 6 - 7 957 91

View Full Product Details