The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163505.1 | NP_001156977.1 | 12 - 13 | 1817 | 64 |
NM_146091.4 | NP_666203.3 | 12 - 13 | 1719 | 64 | |
GenBank mRNA | AK028842.1 | - | 12 - 13 | 1580 | 64 |
AK030660.1 | - | 12 - 13 | 1579 | 64 | |
AK077752.1 | - | 12 - 13 | 1582 | 64 | |
AK147720.1 | - | 12 - 13 | 1677 | 64 | |
AK169476.1 | - | 12 - 13 | 1579 | 64 | |
AK170550.1 | - | 12 - 13 | 3094 | 64 | |
BC002149.1 | - | 9 - 10 | 1109 | 64 | |
BC017138.1 | - | 12 - 13 | 1568 | 64 |