Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Appl2
Entrez Gene ID
216190
Gene Name
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2
Gene Aliases
Dip3b
Chromosome Location
Chr.10: 83600033 - 83648854 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145220.2 NP_660255.1 20 - 21 1936 117
GenBank mRNA AK160075.1 - 20 - 21 1915 117
AK160105.1 - 20 - 21 1912 117
AK168403.1 - 20 - 21 1899 117
AK168748.1 - 20 - 21 1892 117
AK171512.1 - 20 - 21 1892 117
AY113706.1 - 20 - 21 1938 117
BC048906.1 - 20 - 21 1874 117

View Full Product Details