Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmmr
Entrez Gene ID
15366
Gene Name
hyaluronan mediated motility receptor (RHAMM)
Gene Aliases
AA386826, CD168, Rhamm
Chromosome Location
Chr.11: 40701388 - 40733464 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013552.2 NP_038580.2 11 - 12 1338 62
GenBank mRNA AF031932.1 - 11 - 12 1323 62
AF079222.1 - 12 - 13 1287 62
AK143123.1 - 11 - 12 1338 62
AK163410.1 - 11 - 12 1281 62
X64550.1 - 7 - 8 951 62

View Full Product Details