Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Carns1
Entrez Gene ID
107239
Gene Name
carnosine synthase 1
Gene Aliases
AI790298, Atpgd1, mKIAA1394
Chromosome Location
Chr.19: 4164324 - 4175479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134148.2 NP_598909.2 6 - 7 1130 98
GenBank mRNA AK129347.1 - 5 - 6 1346 98

View Full Product Details