Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nrxn2
Entrez Gene ID
18190
Gene Name
neurexin II
Gene Aliases
6430591O13Rik, mKIAA0921
Chromosome Location
Chr.19: 6418738 - 6533217 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205234.1 NP_001192163.1 14 - 15 3370 69
NM_001205235.1 NP_001192164.1 13 - 14 3346 69
NM_020253.3 NP_064649.2 14 - 15 3370 69
GenBank mRNA AK129239.1 - 14 - 15 3343 69
AK147295.1 - 14 - 15 3306 69
AK147665.1 - 14 - 15 3305 69
BC145496.1 - 12 - 13 2993 69
BC150784.1 - 12 - 13 2993 69

View Full Product Details