The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027244.1 | NP_081520.1 | 3 - 4 | 398 | 76 |
GenBank mRNA | AK008201.1 | - | 3 - 4 | 393 | 76 |
BC059729.1 | - | 3 - 4 | 337 | 76 |