Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efhc1
Entrez Gene ID
71877
Gene Name
EF-hand domain (C-terminal) containing 1
Gene Aliases
1700029F22Rik, mRib72-1, myoclonin1
Chromosome Location
Chr.1: 20951551 - 20990841 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027974.1 NP_082250.1 6 - 7 1239 102
GenBank mRNA AK006489.1 - 6 - 7 1178 102
EU520262.1 - 6 - 7 1131 102

View Full Product Details