Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sptbn2
Entrez Gene ID
20743
Gene Name
spectrin beta, non-erythrocytic 2
Gene Aliases
Spnb3, mKIAA0302
Chromosome Location
Chr.19: 4711223 - 4752360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021287.1 NP_067262.1 24 - 25 5427 72
GenBank mRNA AK147671.1 - 24 - 25 5443 72
AK172931.1 - 14 - 15 3736 72
BC048851.1 - 2 - 3 245 72
BC079628.1 - 2 - 3 379 72
BC079860.1 - 24 - 25 5427 72

View Full Product Details