The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007658.3 | NP_031684.3 | 14 - 15 | 1811 | 90 |
GenBank mRNA | AK077609.1 | - | 14 - 15 | 1429 | 90 |
AK077991.1 | - | 14 - 15 | 1813 | 90 | |
BC046296.1 | - | 14 - 15 | 1430 | 90 |