The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025631.3 | NP_079907.2 | 1 - 2 | 78 | 144 |
GenBank mRNA | AK009346.1 | - | 1 - 2 | 51 | 144 |
AK009504.1 | - | 1 - 2 | 53 | 144 | |
AK009619.1 | - | 1 - 2 | 51 | 144 | |
AK009794.1 | - | 1 - 2 | 49 | 144 | |
AK010120.1 | - | 1 - 2 | 49 | 144 | |
AK010154.1 | - | 1 - 2 | 49 | 144 | |
AK010157.1 | - | 1 - 2 | 79 | 144 | |
AK010215.1 | - | 1 - 2 | 49 | 144 | |
AK028156.1 | - | 1 - 2 | 49 | 144 | |
AK028158.1 | - | 1 - 2 | 52 | 144 | |
AK075874.1 | - | 1 - 2 | 49 | 144 | |
AK160516.1 | - | 1 - 2 | 49 | 144 |