Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnah17
Entrez Gene ID
69926
Gene Name
dynein, axonemal, heavy chain 17
Gene Aliases
2810003K23Rik, Dnahc17, Dnahcl1, Gm1178, mKIAA3028
Chromosome Location
Chr.11: 118021723 - 118130961 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167746.1 NP_001161218.1 40 - 41 6215 89

View Full Product Details