The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081240.3 | NP_001074709.1 | 10 - 11 | 2355 | 101 |
GenBank mRNA | AK045315.1 | - | 10 - 11 | 2345 | 101 |
AK154546.1 | - | 10 - 11 | 2299 | 101 | |
AK161023.1 | - | 10 - 11 | 2264 | 101 |