Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Brd7
Entrez Gene ID
26992
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, Ptpn13ip
Chromosome Location
Chr.8: 88330598 - 88362255 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012047.2 NP_036177.1 14 - 15 1850 114
GenBank mRNA AF084259.1 - 14 - 15 1823 114
AK004429.1 - 2 - 3 182 114
AK137117.1 - 1 - 2 1384 114
BC128364.1 - 1 - 2 1150 114
BC152350.1 - 1 - 2 1150 114

View Full Product Details