Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brd7
Entrez Gene ID
26992
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, Ptpn13ip
Chromosome Location
Chr.8: 88330598 - 88362255 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012047.2 NP_036177.1 5 - 6 819 214
GenBank mRNA AF084259.1 - 5 - 6 792 214
AK138934.1 - 5 - 6 823 214

View Full Product Details