Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heatr3
Entrez Gene ID
234549
Gene Name
HEAT repeat containing 3
Gene Aliases
C030036P15Rik
Chromosome Location
Chr.8: 88137885 - 88171952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172757.3 NP_766345.3 3 - 4 538 102
GenBank mRNA AK047782.1 - 3 - 4 569 102
AK052635.1 - 3 - 4 565 102
AK136424.1 - 3 - 4 548 102
AK140522.1 - 3 - 4 556 102
AK163715.1 - 3 - 4 525 102
AK165541.1 - 3 - 4 433 102
BC065060.1 - 3 - 4 538 102

View Full Product Details