The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029467.3 | NP_083743.2 | 2 - 3 | 281 | 75 |
GenBank mRNA | AK016263.1 | - | 2 - 3 | 151 | 75 |
AK076581.1 | - | 2 - 3 | 201 | 75 | |
BC050746.1 | - | - | 205 | 75 |